Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhbg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021375.3 | NP_067350.2 | 6 - 7 | 1002 | 87 | |
GenBank mRNA | AF193808.1 | - | 6 - 7 | 1002 | 87 | |
AK081882.1 | - | 6 - 7 | 1003 | 87 | ||
AY254685.1 | - | 6 - 7 | 990 | 87 | ||
BC029236.1 | - | 6 - 7 | 1020 | 4218889 | 87 | |
FJ387503.1 | - | 1120 | 87 | |||
FJ387504.1 | - | 899 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |