Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uba1y | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011667.2 | NP_035797.1 | 1 - 2 | 101 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |