Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289514.1 | NP_001276443.1 | 2 - 3 | 618 | 67 | |
NM_019636.3 | NP_062610.2 | 3 - 4 | 1199 | 67 | ||
GenBank mRNA | AK122445.1 | - | 3 - 4 | 2008 | 67 | |
AK135892.1 | - | 2 - 3 | 883 | 67 | ||
AK148356.1 | - | 2 - 3 | 574 | 67 | ||
AK154591.1 | - | 4 - 5 | 1235 | 67 | ||
AK159256.1 | - | 3 - 4 | 1195 | 67 | ||
BC059824.1 | - | 4 - 5 | 1564 | 67 | ||
BC117931.1 | - | 2 - 3 | 867 | 40097712 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |