Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289514.1 | NP_001276443.1 | 8 - 9 | 1280 | 62 | |
NM_019636.3 | NP_062610.2 | 9 - 10 | 1861 | 62 | ||
GenBank mRNA | AK122445.1 | - | 9 - 10 | 2670 | 62 | |
AK135665.1 | - | 8 - 9 | 1452 | 62 | ||
AK135892.1 | - | 8 - 9 | 1545 | 62 | ||
AK148356.1 | - | 8 - 9 | 1237 | 62 | ||
AK154591.1 | - | 10 - 11 | 1897 | 62 | ||
AK159256.1 | - | 9 - 10 | 1857 | 62 | ||
BC059824.1 | - | 10 - 11 | 2226 | 62 | ||
BC117931.1 | - | 8 - 9 | 1529 | 40097712 | 62 | |
U33005.1 | - | 9 - 10 | 1817 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |