Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpini1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009250.2 | NP_033276.1 | 1 - 2 | 105 | 79 | |
GenBank mRNA | AJ001700.1 | - | 1 - 2 | 74 | 79 | |
AK032152.1 | - | 1 - 2 | 94 | 79 | ||
AK133770.1 | - | 1 - 2 | 94 | 79 | ||
AK164355.1 | - | 1 - 2 | 60 | 79 | ||
BC006776.1 | - | 1 - 2 | 70 | 3588856 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |