Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc9a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177909.5 | NP_808577.3 | 2 - 3 | 546 | 100 | |
GenBank mRNA | AK037058.1 | - | 2 - 3 | 530 | 100 | |
AK156494.1 | - | 2 - 3 | 507 | 100 | ||
AK172687.1 | - | 2 - 3 | 518 | 100 | ||
BC099942.1 | - | 2 - 3 | 520 | 30103308 | 100 | |
BC153825.1 | - | 2 - 3 | 524 | 40093280 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |