Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009355.2 | NP_033381.1 | 2 - 3 | 356 | 75 | |
GenBank mRNA | AB008910.1 | - | 2 - 3 | 356 | 75 | |
BC049616.1 | - | 2 - 3 | 351 | 6773970 | 75 | |
BC115665.2 | - | 2 - 3 | 355 | 40103370 | 75 | |
BC115666.2 | - | 2 - 3 | 355 | 40103373 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |