Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pi15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053191.2 | NP_444421.2 | 1 - 2 | 145 | 93 | |
GenBank mRNA | AF329196.1 | - | 1 - 2 | 65 | 93 | |
AK030960.1 | - | 1 - 2 | 146 | 93 | ||
AK040961.1 | - | 1 - 2 | 144 | 93 | ||
BC116705.1 | - | 1 - 2 | 41 | 40102877 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |