Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010267.3 | NP_034397.1 | 3 - 4 | 585 | 85 | |
GenBank mRNA | AK020988.1 | - | 3 - 4 | 569 | 85 | |
AK045460.1 | - | 3 - 4 | 585 | 85 | ||
AK083814.1 | - | 3 - 4 | 586 | 85 | ||
AK139057.1 | - | 3 - 4 | 428 | 85 | ||
AK149281.1 | - | 3 - 4 | 596 | 85 | ||
BC048177.1 | - | 3 - 4 | 561 | 6825756 | 85 | |
BC051135.1 | - | 3 - 4 | 550 | 6532839 | 85 | |
BC060212.1 | - | 3 - 4 | 558 | 85 | ||
Y17850.1 | - | 3 - 4 | 573 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |