Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190451.1 | NP_001177380.1 | 3 - 4 | 628 | 128 | |
NM_007833.5 | NP_031859.1 | 3 - 4 | 536 | 128 | ||
GenBank mRNA | AK135287.1 | - | 3 - 4 | 520 | 128 | |
AK145905.1 | - | 3 - 4 | 518 | 128 | ||
AK147020.1 | - | 3 - 4 | 520 | 128 | ||
AK161788.1 | - | 3 - 4 | 518 | 128 | ||
AK165408.1 | - | 3 - 4 | 518 | 128 | ||
BC060126.1 | - | 2 - 3 | 349 | 128 | ||
BC132521.1 | - | 3 - 4 | 387 | 40130798 | 128 | |
BC138564.1 | - | 3 - 4 | 387 | 8861584 | 128 | |
X53929.1 | - | 3 - 4 | 545 | 128 | ||
Y11757.1 | - | 2 - 3 | 234 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |