Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190451.1 | NP_001177380.1 | 6 - 7 | 1039 | 62 | |
NM_007833.5 | NP_031859.1 | 6 - 7 | 947 | 62 | ||
GenBank mRNA | AK135287.1 | - | 6 - 7 | 931 | 62 | |
AK145905.1 | - | 6 - 7 | 929 | 62 | ||
AK147020.1 | - | 6 - 7 | 931 | 62 | ||
AK161788.1 | - | 6 - 7 | 929 | 62 | ||
AK165408.1 | - | 6 - 7 | 928 | 62 | ||
BC132521.1 | - | 6 - 7 | 798 | 40130798 | 62 | |
BC138564.1 | - | 6 - 7 | 798 | 8861584 | 62 | |
X53929.1 | - | 6 - 7 | 956 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |