Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100000129.mm.1 |
NM_009826.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000129.mm.1 |
NM_009826.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000129.mm.1 |
TC0100000021.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100000138.mm.1 |
NM_009826.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000138.mm.1 |
NM_009826.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000138.mm.1 |
TC0100000021.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100000145.mm.1 |
NM_009826.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000145.mm.1 |
NM_009826.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000145.mm.1 |
TC0100000021.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100000128.mm.1 |
NM_009826.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000128.mm.1 |
NM_009826.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000128.mm.1 |
TC0100000021.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100000136.mm.1 |
NM_009826.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000136.mm.1 |
NM_009826.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000136.mm.1 |
TC0100000021.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100000137.mm.1 |
NM_009826.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000137.mm.1 |
NM_009826.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000137.mm.1 |
TC0100000021.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100000143.mm.1 |
NM_009826.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000143.mm.1 |
NM_009826.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000143.mm.1 |
TC0100000021.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100000134.mm.1 |
NM_009826.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000134.mm.1 |
NM_009826.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000134.mm.1 |
TC0100000021.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100000141.mm.1 |
NM_009826.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000141.mm.1 |
NM_009826.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000141.mm.1 |
TC0100000021.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100000117.mm.1 |
NM_009826.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000117.mm.1 |
NM_009826.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000117.mm.1 |
TC0100000021.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100000125.mm.1 |
NM_009826.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000125.mm.1 |
NM_009826.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000125.mm.1 |
TC0100000021.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100000130.mm.1 |
NM_009826.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000130.mm.1 |
NM_009826.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000130.mm.1 |
TC0100000021.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100000131.mm.1 |
NM_009826.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000131.mm.1 |
NM_009826.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000131.mm.1 |
TC0100000021.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100000142.mm.1 |
NM_009826.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000142.mm.1 |
NM_009826.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000142.mm.1 |
TC0100000021.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100000126.mm.1 |
NM_009826.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000126.mm.1 |
NM_009826.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000126.mm.1 |
TC0100000021.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100000132.mm.1 |
NM_009826.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000132.mm.1 |
NM_009826.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000132.mm.1 |
TC0100000021.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100000139.mm.1 |
NM_009826.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000139.mm.1 |
NM_009826.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000139.mm.1 |
TC0100000021.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100000121.mm.1 |
NM_009826.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000121.mm.1 |
NM_009826.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000121.mm.1 |
TC0100000021.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100000120.mm.1 |
NM_009826.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000120.mm.1 |
NM_009826.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000120.mm.1 |
TC0100000021.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100000140.mm.1 |
NM_009826.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000140.mm.1 |
NM_009826.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000140.mm.1 |
TC0100000021.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100000144.mm.1 |
NM_009826.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000144.mm.1 |
NM_009826.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000144.mm.1 |
TC0100000021.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100000123.mm.1 |
NM_009826.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000123.mm.1 |
NM_009826.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000123.mm.1 |
TC0100000021.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100000127.mm.1 |
NM_009826.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000127.mm.1 |
NM_009826.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000127.mm.1 |
TC0100000021.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100000133.mm.1 |
NM_009826.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000133.mm.1 |
NM_009826.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000133.mm.1 |
TC0100000021.mm.1 |
MTA TC |