Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000008281.mm.1 |
NM_029166.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008281.mm.1 |
NM_029166.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008281.mm.1 |
TC1000001089.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000008277.mm.1 |
NM_029166.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008277.mm.1 |
NM_029166.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008277.mm.1 |
TC1000001089.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000008284.mm.1 |
NM_029166.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008284.mm.1 |
NM_029166.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008284.mm.1 |
TC1000001089.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000008283.mm.1 |
NM_029166.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008283.mm.1 |
NM_029166.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008283.mm.1 |
TC1000001089.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000008282.mm.1 |
NM_029166.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008282.mm.1 |
NM_029166.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008282.mm.1 |
TC1000001089.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000008276.mm.1 |
NM_029166.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008276.mm.1 |
NM_029166.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008276.mm.1 |
TC1000001089.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000008278.mm.1 |
NM_029166.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008278.mm.1 |
NM_029166.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008278.mm.1 |
TC1000001089.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000008274.mm.1 |
NM_029166.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008274.mm.1 |
NM_029166.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008274.mm.1 |
TC1000001089.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000008275.mm.1 |
NM_029166.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008275.mm.1 |
NM_029166.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008275.mm.1 |
TC1000001089.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000008285.mm.1 |
NM_029166.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008285.mm.1 |
NM_029166.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008285.mm.1 |
TC1000001089.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000008271.mm.1 |
NM_029166.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008271.mm.1 |
NM_029166.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008271.mm.1 |
TC1000001089.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000008272.mm.1 |
NM_029166.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008272.mm.1 |
NM_029166.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008272.mm.1 |
TC1000001089.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000008280.mm.1 |
NM_029166.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008280.mm.1 |
NM_029166.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008280.mm.1 |
TC1000001089.mm.1 |
MTA TC |