Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ric8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013441.2 | NP_001013459.1 | 6 - 7 | 1295 | 119 | |
NM_183172.2 | NP_898995.1 | 6 - 7 | 1295 | 119 | ||
GenBank mRNA | AK132369.1 | - | 6 - 7 | 1199 | 119 | |
AK137531.1 | - | 5 - 6 | 1093 | 119 | ||
AY940666.1 | - | 6 - 7 | 1264 | 119 | ||
AY940667.1 | - | 6 - 7 | 1252 | 119 | ||
BC051080.1 | - | 6 - 7 | 1292 | 6490136 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |