Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028882.4 | NP_083158.3 | 6 - 7 | 1136 | 75 | |
GenBank mRNA | AK028900.1 | - | 6 - 7 | 918 | 75 | |
AK031704.1 | - | 6 - 7 | 748 | 75 | ||
AK051165.1 | - | 6 - 7 | 797 | 75 | ||
AK052671.1 | - | 6 - 7 | 1137 | 75 | ||
AK134412.1 | - | 6 - 7 | 772 | 75 | ||
AK153839.1 | - | 6 - 7 | 797 | 75 | ||
BC138131.1 | - | 6 - 7 | 679 | 8861150 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |