Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Heph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159627.1 | NP_001153099.1 | 5 - 6 | 1123 | 91 | |
NM_001159628.1 | NP_001153100.1 | 5 - 6 | 1131 | 91 | ||
NM_010417.2 | NP_034547.2 | 5 - 6 | 1131 | 91 | ||
NM_181273.4 | NP_851790.1 | 5 - 6 | 1123 | 91 | ||
GenBank mRNA | AF082567.1 | - | 5 - 6 | 1243 | 91 | |
AK032895.1 | - | 5 - 6 | 1043 | 91 | ||
AK081330.1 | - | 5 - 6 | 1099 | 91 | ||
AK086923.1 | - | 5 - 6 | 1042 | 91 | ||
AK129194.1 | - | 5 - 6 | 1079 | 91 | ||
BC048237.1 | - | 5 - 6 | 971 | 6474714 | 91 | |
BC054442.1 | - | 5 - 6 | 971 | 6474713 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |