Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Awat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081136.1 | NP_001074605.1 | 2 - 3 | 278 | 70 | |
GenBank mRNA | AK137763.1 | - | 2 - 3 | 275 | 70 | |
BC147680.1 | - | 2 - 3 | 249 | 9056653 | 70 | |
BC147683.1 | - | 2 - 3 | 249 | 9056656 | 70 | |
BC147829.1 | - | 2 - 3 | 207 | 9087546 | 70 | |
BC147832.1 | - | 2 - 3 | 207 | 9087549 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |