Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177715.4 | NP_808383.3 | 1 | 2065 | 71 | |
GenBank mRNA | AK035535.1 | - | 1 | 2067 | 71 | |
AK049420.1 | - | 1 | 190 | 71 | ||
AK172581.1 | - | 1 | 2066 | 71 | ||
AK173260.1 | - | 1 | 1570 | 71 | ||
BC116647.1 | - | 2 | 1452 | 71 | ||
BC116648.1 | - | 2 | 1452 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |