Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177352.4 | NP_796326.1 | 9 - 10 | 853 | 79 | |
GenBank mRNA | AK049275.1 | - | 9 - 10 | 854 | 79 | |
AK076293.1 | - | 9 - 10 | 882 | 79 | ||
AK160477.1 | - | 9 - 10 | 908 | 79 | ||
AK162119.1 | - | 9 - 10 | 854 | 79 | ||
AK163031.1 | - | 9 - 10 | 849 | 79 | ||
AK167217.1 | - | 9 - 10 | 873 | 79 | ||
BC116302.1 | - | 9 - 10 | 850 | 40095102 | 79 | |
BC116303.1 | - | 8 - 9 | 756 | 40095106 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |