Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hs6st3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK139324.1 | - | 1 | 1346 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |