Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008448.3 | NP_032474.2 | 7 - 8 | 1001 | 124 | |
GenBank mRNA | AK133535.1 | - | 7 - 8 | 998 | 124 | |
AK136307.1 | - | 7 - 8 | 998 | 124 | ||
BC013248.1 | - | 7 - 8 | 957 | 124 | ||
BC013817.1 | - | 7 - 8 | 967 | 124 | ||
BC025864.1 | - | 7 - 8 | 970 | 124 | ||
BC069920.1 | - | 7 - 8 | 977 | 124 | ||
BC090841.1 | - | 7 - 8 | 977 | 30543821 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |