Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008549.2 | NP_032575.2 | 14 - 15 | 2413 | 67 | |
GenBank mRNA | BC138371.1 | - | 14 - 15 | 2398 | 8861391 | 67 |
BC138372.1 | - | 14 - 15 | 2398 | 8861392 | 67 | |
X61172.1 | - | 14 - 15 | 2413 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |