Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000009789.mm.1 |
NM_178278.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009789.mm.1 |
NM_178278.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009789.mm.1 |
TC1000001307.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000009776.mm.1 |
NM_178278.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009776.mm.1 |
NM_178278.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009776.mm.1 |
TC1000001307.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000009782.mm.1 |
NM_178278.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009782.mm.1 |
NM_178278.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009782.mm.1 |
TC1000001307.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000009788.mm.1 |
NM_178278.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009788.mm.1 |
NM_178278.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009788.mm.1 |
TC1000001307.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000009785.mm.1 |
NM_178278.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009785.mm.1 |
NM_178278.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009785.mm.1 |
TC1000001307.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000009790.mm.1 |
NM_178278.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009790.mm.1 |
NM_178278.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009790.mm.1 |
TC1000001307.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000009780.mm.1 |
NM_178278.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009780.mm.1 |
NM_178278.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009780.mm.1 |
TC1000001307.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000009784.mm.1 |
NM_178278.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009784.mm.1 |
NM_178278.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009784.mm.1 |
TC1000001307.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000009787.mm.1 |
NM_178278.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009787.mm.1 |
NM_178278.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009787.mm.1 |
TC1000001307.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000009791.mm.1 |
NM_178278.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009791.mm.1 |
NM_178278.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009791.mm.1 |
TC1000001307.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000009781.mm.1 |
NM_178278.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009781.mm.1 |
NM_178278.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009781.mm.1 |
TC1000001307.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000009786.mm.1 |
NM_178278.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009786.mm.1 |
NM_178278.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009786.mm.1 |
TC1000001307.mm.1 |
MTA TC |