Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vprbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015507.2 | NP_001015507.1 | 19 - 20 | 4046 | 106 | |
GenBank mRNA | AK029372.1 | - | 2 - 3 | 120 | 106 | |
AK122372.1 | - | 19 - 20 | 4009 | 106 | ||
BC065119.1 | - | 19 - 20 | 4046 | 6853519 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |