Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vprbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015507.2 | NP_001015507.1 | 5 - 6 | 499 | 69 | |
GenBank mRNA | AK122372.1 | - | 5 - 6 | 462 | 69 | |
BC065119.1 | - | 5 - 6 | 499 | 6853519 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |