Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100016120.mm.1 |
NM_172841.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100016120.mm.1 |
NM_172841.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100016120.mm.1 |
TC0100001967.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100016121.mm.1 |
NM_172841.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100016121.mm.1 |
NM_172841.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100016121.mm.1 |
TC0100001967.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100016123.mm.1 |
NM_172841.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100016123.mm.1 |
NM_172841.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100016123.mm.1 |
TC0100001967.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100016122.mm.1 |
NM_172841.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100016122.mm.1 |
NM_172841.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100016122.mm.1 |
TC0100001967.mm.1 |
MTA TC |