Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sulf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198565.1 | NP_001185494.1 | 5 - 6 | 892 | 88 | |
NM_001198566.1 | NP_001185495.1 | 4 - 5 | 797 | 88 | ||
NM_172294.1 | NP_758498.1 | 5 - 6 | 876 | 88 | ||
GenBank mRNA | AK028285.1 | - | 4 - 5 | 696 | 88 | |
AK045002.1 | - | 5 - 6 | 791 | 88 | ||
AK078756.1 | - | 5 - 6 | 881 | 88 | ||
AK129278.1 | - | 4 - 5 | 698 | 88 | ||
AY101178.1 | - | 5 - 6 | 901 | 88 | ||
BC034547.1 | - | 5 - 6 | 876 | 4500954 | 88 | |
BC049276.1 | - | 5 - 6 | 876 | 4500954 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |