Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il1r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123382.1 | NP_001116854.1 | 3 - 4 | 602 | 99 | |
NM_008362.2 | NP_032388.1 | 3 - 4 | 533 | 99 | ||
GenBank mRNA | AK048550.1 | - | 3 - 4 | 520 | 99 | |
AK054137.1 | - | 3 - 4 | 603 | 99 | ||
AK166301.1 | - | 3 - 4 | 544 | 99 | ||
BC109135.2 | - | 3 - 4 | 439 | 40039321 | 99 | |
M20658.1 | - | 3 - 4 | 533 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |