Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177606.4 | NP_808274.2 | 28 - 29 | 4232 | 145 | |
GenBank mRNA | AK029252.1 | - | 28 - 29 | 4234 | 145 | |
AK086794.1 | - | 8 - 9 | 1062 | 145 | ||
AK173337.1 | - | 25 - 26 | 3887 | 145 | ||
BC125583.1 | - | 28 - 29 | 4197 | 40130098 | 145 | |
BC137804.1 | - | 28 - 29 | 4194 | 8860823 | 145 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |