Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161837.1 | NP_001155309.1 | 6 - 7 | 972 | 97 | |
NM_001161838.1 | NP_001155310.1 | 3 - 4 | 929 | 97 | ||
NM_001161839.1 | NP_001155311.1 | 4 - 5 | 620 | 97 | ||
NM_001161840.1 | NP_001155312.1 | 4 - 5 | 812 | 97 | ||
NM_011217.2 | NP_035347.1 | 7 - 8 | 1578 | 97 | ||
GenBank mRNA | AF041866.1 | - | 4 - 5 | 737 | 97 | |
AF129509.1 | - | 3 - 4 | 854 | 97 | ||
AK028054.1 | - | 7 - 8 | 1579 | 97 | ||
BC119230.1 | - | 7 - 8 | 1511 | 8733979 | 97 | |
BC120547.1 | - | 7 - 8 | 1511 | 40129470 | 97 | |
D31898.1 | - | 7 - 8 | 1541 | 97 | ||
Z30313.1 | - | 6 - 7 | 967 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |