Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008446.2 | NP_032472.2 | 2 - 3 | 635 | 96 | |
GenBank mRNA | AK084837.1 | - | 2 - 3 | 612 | 96 | |
AK135179.1 | - | 2 - 3 | 583 | 96 | ||
AK135430.1 | - | 2 - 3 | 129 | 96 | ||
AK171938.1 | - | 2 - 3 | 549 | 96 | ||
BC050946.1 | - | 2 - 3 | 573 | 6822283 | 96 | |
BC054537.1 | - | 2 - 3 | 557 | 96 | ||
D12646.1 | - | 2 - 3 | 962 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |