Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100017718.mm.1 |
NM_173870.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100017718.mm.1 |
TC0100002204.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100017728.mm.1 |
NM_173870.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100017728.mm.1 |
TC0100002204.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100017725.mm.1 |
NM_173870.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100017725.mm.1 |
TC0100002204.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100017713.mm.1 |
NM_173870.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100017713.mm.1 |
TC0100002204.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100017714.mm.1 |
NM_173870.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100017714.mm.1 |
TC0100002204.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100017724.mm.1 |
NM_173870.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100017724.mm.1 |
TC0100002204.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100017729.mm.1 |
NM_173870.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100017729.mm.1 |
TC0100002204.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100017716.mm.1 |
NM_173870.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100017716.mm.1 |
TC0100002204.mm.1 |
MTA TC |