Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198303.2 | NP_938045.2 | 16 - 17 | 2859 | 66 | |
GenBank mRNA | BC103806.1 | - | 15 - 16 | 3612 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |