Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epcam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008532.2 | NP_032558.2 | 6 - 7 | 1011 | 116 | |
GenBank mRNA | AK133510.1 | - | 5 - 6 | 693 | 116 | |
AK145752.1 | - | 5 - 6 | 587 | 116 | ||
AK167182.1 | - | 5 - 6 | 656 | 116 | ||
BC005618.1 | - | 5 - 6 | 652 | 3711069 | 116 | |
BC094465.1 | - | 5 - 6 | 677 | 4948935 | 116 | |
M76124.1 | - | 5 - 6 | 660 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |