Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rapgef5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175930.5 | NP_787126.3 | 25 - 26 | 2851 | 143 | |
GenBank mRNA | AK029995.1 | - | 25 - 26 | 2852 | 143 | |
AK030016.1 | - | 25 - 26 | 2805 | 143 | ||
AK083591.1 | - | 15 - 16 | 1791 | 143 | ||
BC046627.1 | - | 13 - 14 | 1572 | 6494175 | 143 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |