Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tut1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_197993.2 | NP_932110.1 | 3 - 4 | 677 | 93 | |
GenBank mRNA | AK138418.1 | - | 3 - 4 | 597 | 93 | |
BC023900.1 | - | 3 - 4 | 654 | 5321265 | 93 | |
BC025499.1 | - | 3 - 4 | 701 | 5320398 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |