Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd13d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026720.2 | NP_080996.2 | 8 - 9 | 1046 | 70 | |
GenBank mRNA | AK089186.1 | - | 8 - 9 | 1039 | 70 | |
BC058982.1 | - | 8 - 9 | 957 | 5685059 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |