Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135198.1 | NP_001128670.1 | 3 - 4 | 405 | 92 | |
GenBank mRNA | BC058624.1 | - | 3 - 4 | 410 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |