Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135198.1 | NP_001128670.1 | 4 - 5 | 457 | 89 | |
GenBank mRNA | BC058624.1 | - | 4 - 5 | 462 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |