Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039692.1 | NP_001034781.1 | 10 - 11 | 1913 | 90 | |
NM_029277.2 | NP_083553.2 | 9 - 10 | 1838 | 90 | ||
GenBank mRNA | AK031648.1 | - | 10 - 11 | 1757 | 90 | |
AK037784.1 | - | 3 - 4 | 1477 | 90 | ||
AK077063.1 | - | 10 - 11 | 1914 | 90 | ||
BC024633.1 | - | 5 - 6 | 325 | 90 | ||
BC037608.1 | - | 9 - 10 | 1653 | 90 | ||
BC141184.1 | - | 9 - 10 | 1647 | 9055796 | 90 | |
BC145506.1 | - | 9 - 10 | 1646 | 9054069 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |