Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039692.1 | NP_001034781.1 | 14 - 15 | 2342 | 83 | |
NM_029277.2 | NP_083553.2 | 13 - 14 | 2267 | 83 | ||
GenBank mRNA | AK031648.1 | - | 14 - 15 | 2186 | 83 | |
AK077063.1 | - | 14 - 15 | 2343 | 83 | ||
BC024633.1 | - | 9 - 10 | 754 | 83 | ||
BC037608.1 | - | 13 - 14 | 2082 | 83 | ||
BC141184.1 | - | 13 - 14 | 2076 | 9055796 | 83 | |
BC145506.1 | - | 13 - 14 | 2075 | 9054069 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |