Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011678.2 | NP_035808.2 | 14 - 15 | 1972 | 79 | |
GenBank mRNA | AK083423.1 | - | 4 - 5 | 467 | 79 | |
AK143582.1 | - | 14 - 15 | 1907 | 79 | ||
AK149964.1 | - | 14 - 15 | 1930 | 79 | ||
AK164245.1 | - | 5 - 6 | 747 | 79 | ||
AK169372.1 | - | 14 - 15 | 1951 | 79 | ||
AK169933.1 | - | 14 - 15 | 1942 | 79 | ||
AK171271.1 | - | 14 - 15 | 1942 | 79 | ||
AK171633.1 | - | 14 - 15 | 1925 | 79 | ||
AK220512.1 | - | 14 - 15 | 1929 | 79 | ||
BC011341.1 | - | 13 - 14 | 1749 | 3661165 | 79 | |
BC066180.1 | - | 14 - 15 | 1882 | 79 | ||
BC066865.1 | - | 14 - 15 | 1882 | 79 | ||
L00681.1 | - | 14 - 15 | 1935 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |