Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pawr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054056.2 | NP_473397.1 | 1 - 2 | 60 | 93 | |
GenBank mRNA | BC027693.1 | - | 1 - 2 | 95 | 93 | |
BC050234.1 | - | 1 - 2 | 211 | 93 | ||
DQ363525.1 | - | 1 - 2 | 60 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |