Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plscr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178711.3 | NP_848826.1 | 5 - 6 | 954 | 138 | |
GenBank mRNA | AK035878.1 | - | 5 - 6 | 939 | 138 | |
AK035919.1 | - | 5 - 6 | 955 | 138 | ||
AK045372.1 | - | 5 - 6 | 955 | 138 | ||
AK054258.1 | - | 5 - 6 | 512 | 138 | ||
AK082330.1 | - | 4 - 5 | 397 | 138 | ||
AK141580.1 | - | 480 | 138 | |||
AK165919.1 | - | 5 - 6 | 955 | 138 | ||
AK166080.1 | - | 5 - 6 | 446 | 138 | ||
BC052067.1 | - | 5 - 6 | 508 | 5708712 | 138 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |