Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900009258.mm.1 |
NM_001142916.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009258.mm.1 |
NM_001142916.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009258.mm.1 |
NM_011961.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009258.mm.1 |
NM_011961.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009258.mm.1 |
TC0900001175.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900009261.mm.1 |
NM_001142916.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009261.mm.1 |
NM_001142916.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009261.mm.1 |
NM_011961.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009261.mm.1 |
NM_011961.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009261.mm.1 |
TC0900001175.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900009257.mm.1 |
NM_001142916.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009257.mm.1 |
NM_001142916.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009257.mm.1 |
NM_011961.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009257.mm.1 |
NM_011961.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009257.mm.1 |
TC0900001175.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900009259.mm.1 |
NM_001142916.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009259.mm.1 |
NM_001142916.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009259.mm.1 |
NM_011961.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009259.mm.1 |
NM_011961.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009259.mm.1 |
TC0900001175.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900009260.mm.1 |
NM_001142916.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009260.mm.1 |
NM_001142916.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009260.mm.1 |
NM_011961.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009260.mm.1 |
NM_011961.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009260.mm.1 |
TC0900001175.mm.1 |
MTA TC |