Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rev1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019570.3 | NP_062516.2 | 2 - 3 | 270 | 81 | |
GenBank mRNA | AB057418.1 | - | 1 - 2 | 59 | 81 | |
AF179302.1 | - | 1 - 2 | 59 | 81 | ||
AK048838.1 | - | 2 - 3 | 275 | 81 | ||
BC058093.1 | - | 2 - 3 | 270 | 6827108 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |