Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rev1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019570.3 | NP_062516.2 | 6 - 7 | 1418 | 63 | |
GenBank mRNA | AB057418.1 | - | 5 - 6 | 1207 | 63 | |
AF179302.1 | - | 5 - 6 | 1207 | 63 | ||
AK048838.1 | - | 6 - 7 | 1423 | 63 | ||
BC006721.1 | - | 1 - 2 | 162 | 63 | ||
BC058093.1 | - | 6 - 7 | 1418 | 6827108 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |