Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cebpz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024806.2 | NP_001019977.1 | 13 - 14 | 2919 | 111 | |
GenBank mRNA | AK145374.1 | - | 13 - 14 | 2901 | 111 | |
AK155990.1 | - | 13 - 14 | 2902 | 111 | ||
AK158771.1 | - | 13 - 14 | 2896 | 111 | ||
AK159081.1 | - | 13 - 14 | 2916 | 111 | ||
U19891.1 | - | 13 - 14 | 2889 | 111 | ||
U19892.1 | - | 13 - 14 | 2603 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |