Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146107.2 | NP_666219.1 | 2 - 3 | 187 | 74 | |
GenBank mRNA | AK078508.1 | - | 2 - 3 | 203 | 74 | |
AK088839.1 | - | 2 - 3 | 220 | 74 | ||
BC023020.1 | - | 2 - 3 | 187 | 5375425 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |