Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bfsp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002896.2 | NP_001002896.1 | 1 - 2 | 601 | 54 | |
GenBank mRNA | AK134810.1 | - | 1 - 2 | 533 | 54 | |
BC068172.1 | - | 1 - 2 | 601 | 30550909 | 54 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |